MilliporeSigma
  • Rhodium(III) and iridium(III) complexes with 1,2-naphthoquinone-1-oximate as a bidentate ligand: synthesis, structure, and biological activity.
Rhodium(III) and iridium(III) complexes with 1,2-naphthoquinone-1-oximate as a bidentate ligand: synthesis, structure, and biological activity.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2010-01-22)
Stefan Wirth, Christoph J Rohbogner, Marcin Cieslak, Julia Kazmierczak-Baranska, Stefan Donevski, Barbara Nawrot, Ingo-Peter Lorenz
ABSTRACT
The synthesis and characterization of three novel iridium(III) complexes and one rhodium(III) complex with 1-nitroso-2-naphthol (3) chelating as a 1,2-naphthoquinone-1-oximato ligand are described. The reaction of mu(2)-halogenido-bridged dimers [(eta(5)-C(5)Me(5))IrX(2)](2) [X is Cl (1a), Br (1b), I (1c)] and [(eta(5)-C(5)Me(5))RhCl(2)](2) (2a) with 3 in CH(2)Cl(2) yields the mononuclear complexes (eta(5)-C(5)Me(5))IrX(eta(2)-C(10)H(6)N(2)O) (4a, 4b, 4c) and (eta(5)-C(5)Me(5))RhCl(eta(2)-C(10)H(6)N(2)O) (5a). All compounds were characterized by their (1)H and (13)C NMR, IR, and mass spectra, UV/vis spectra were recorded for 4a and 5a. The X-ray structure analyses revealed a pseudo-octahedral "piano-stool" configuration for the metals with bidentate coordination through oximato-N and naphthoquinone-O, forming a nearly planar five-membered metallacycle. The metal complexes 4a and 5a were evaluated in respect to their cytotoxicity and binding affinity toward double-stranded DNA. As determined in the 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assay, both exerted a much stronger cytotoxic effect toward HeLa and HL60 cancer cell lines than did cisplatin. The remarkable cytotoxicity of the compounds tested may be attributed to necrosis, rather than to apoptosis, as it is evidenced by the caspase-3/7 activation assay. No clear evidence was found for interaction with double-stranded DNA. The melting experiments showed no significant differences between thermodynamic parameters of intact DNA and DNA incubated with 3, 4a, or 5a, although these derivatives altered DNA recognition by the BamHI restriction enzyme. Therefore, the screened iridium and rhodium complexes 4a and 5a may still be interesting as potential anticancer drugs owing to their high cytotoxicity toward cancer cell lines, whereas they do not modify DNA in a way similar to that of cisplatin.