MilliporeSigma
  • Pyridine-functionalized MCM-41 as an efficient and recoverable catalyst for the synthesis of pyran annulated heterocyclic systems.
Pyridine-functionalized MCM-41 as an efficient and recoverable catalyst for the synthesis of pyran annulated heterocyclic systems.
Chemical & pharmaceutical bulletin (2010-02-02)
Ahmad Shaabani, Mostafa Mohammadpour Amini, Sabrieh Ghasemi, Rahim Ghadari, Ali Hossein Rezayan, Yousef Fazaeli, Shahzad Feizi
ABSTRACT
Pyridine-functionalized MCM-41 catalyzed reactions between tetracyanoethylene and various activated CH-acid compounds are described. These reactions afford the corresponding pyran annulated heterocyclic ring systems in high yields at room temperature within a few minutes. The work-up procedure is very simple and the products do not require further purification. The catalyst can be recycled and reused for several times without observable loss of performance.
Sigma-Aldrich
Tetracyanoethylene, 96%