MilliporeSigma
  • Regioselective carboxylation of catechol by 3,4-dihydroxybenzoate decarboxylase of Enterobacter cloacae P.
Regioselective carboxylation of catechol by 3,4-dihydroxybenzoate decarboxylase of Enterobacter cloacae P.
Biotechnology letters (2010-02-05)
Toyokazu Yoshida, Yuki Inami, Tsuyoshi Matsui, Toru Nagasawa
ABSTRACT
3,4-Dihydroxybenzoate decarboxylase in Enterobacter cloacae P241 was induced by adding 3,4-dihydroxybenzoic acid, 3-hydroxybenzoic acid, 3,4,5-trihydroxybenzoic acid or 4-acetamidobenzoic acid to the culture medium. After stabilizing the enzyme activity by adding 5 mM dithiothreitol and 20 mM Na(2)S(2)O(3) to a cell-free extract, catechol at 50 mM was carboxylated in the presence of 3 M KHCO(3) to 3,4-dihydroxybenzoic acid with a molar conversion ratio of 28% after 14 h at 30 degrees C.
Sigma-Aldrich
4-Acetamidobenzoic acid, 98%
Sigma-Aldrich
Potassium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Potassium bicarbonate, ACS reagent, 99.7%, powder, crystals or granules
Sigma-Aldrich
Potassium bicarbonate, ≥99.95% trace metals basis, 99.7-100.5% dry basis
Sigma-Aldrich
Potassium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, E501, 99.5-101.0% (acidimetric)
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%