MilliporeSigma
Effect of hypnotic drugs on body balance and standing steadiness.
Sleep medicine reviews (2010-02-23)
Monique A J Mets, Edmund R Volkerts, Berend Olivier, Joris C Verster
ABSTRACT
Disturbed body balance and standing steadiness are problematic for those who wake up at night or in the morning after using hypnotic drugs. As a result, falls and hip fractures are frequently reported in patients using sleep medication. A literature search was performed to identify double blind, placebo-controlled clinical trials that examined body balance and standing steadiness. Drugs that were searched were nitrazepam, triazolam, lorazepam, temazepam, loprazolam, flunitrazepam, flurazepam, and the Z-drugs zopiclone, zolpidem and zaleplon. A total of 57 studies were eligible for inclusion. Results showed that both benzodiazepine hypnotics and the Z-drugs significantly impair body balance and standing steadiness after single dose administration. Impairments correlate significantly with blood plasma levels and are greatest at peak plasma concentrations, but are sometimes still present upon awakening. Balance problems were dose-related and most pronounced in elderly. Co-administration of alcohol aggravated the impairment. After repeated daily use of hypnotic drugs partial tolerance develops to the impairing effects on standing steadiness. Single dose administration of benzodiazepine hypnotics and Z-drugs significantly impair body balance in a dose-dependent manner. Zolpidem and zopiclone produced similar significant impairment as benzodiazepine hypnotics. Zaleplon significantly impaired balance up to 2 h after intake. Partial tolerance develops after repeated daily use. In conclusion, patients should be warned about the possible risk of imbalance and falls due to the use of sleep medication.
Product Description
Supelco
Zaleplon solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®