MilliporeSigma
  • Quantum chemical DFT study of 4-azatricyclo [5.2.2.0(2,6)] undecane-3,5,8-trione.
Quantum chemical DFT study of 4-azatricyclo [5.2.2.0(2,6)] undecane-3,5,8-trione.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2010-03-17)
C Yohannan Panicker, Hema Tresa Varghese, K Madhusoodanan Pillai, Y Sheena Mary, K Raju, T K Manojkumar, Anna Bielenica, Christian Van Alsenoy
ABSTRACT
Fourier-transform-Raman and infrared spectrum of 4-azatricyclo [5.2.2.0(2,6)] undecane-3,5,8-trione were recorded and analyzed. The vibrational wavenumbers were examined theoretically using the Gaussian03 set of quantum chemistry codes, and the normal modes were assigned by potential energy distribution (PED) calculations. The first hyperpolarizability, predicted infrared intensities and Raman activities are reported. The calculated first hyperpolarizability is comparable with reported values of similar structures which makes this compound an attractive object for future studies of nonlinear optics. Optimized geometrical parameters of the compound are in agreement with similar reported structures. The red shift of the NH stretching wavenumber in the infrared spectrum from the computational wavenumber indicates the weakening of NH bond.
Sigma-Aldrich
Undecane, ≥99%
Sigma-Aldrich
Undecane, ≥99%
Supelco
Undecane, analytical standard