MilliporeSigma
  • Synthesis, cytotoxicity and antileishmanial activity of some N-(2-(indol-3-yl)ethyl)-7-chloroquinolin-4-amines.
Synthesis, cytotoxicity and antileishmanial activity of some N-(2-(indol-3-yl)ethyl)-7-chloroquinolin-4-amines.
Chemical biology & drug design (2010-03-27)
Elaine S Coimbra, Rafael Carvalhaes, Richard M Grazul, Patricia A Machado, Marcos V N De Souza, Adilson D Da Silva
ABSTRACT
We report herein the condensation of 4,7-dichloroquinoline (1) with tryptamine (2) and D-tryptophan methyl ester (3). Hydrolysis of the methyl ester adduct (5) yielded the free acid (6). The compounds were evaluated in vitro for activity against four different species of Leishmania promastigote forms and for cytotoxic activity against Kb and Vero cells. Compound (5) showed good activity against the Leishmania species tested, while all three compounds displayed moderate activity in both Kb and Vero cells.
Sigma-Aldrich
4,7-Dichloroquinoline, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
4,7-Dichloroquinoline, ≥99%
Pricing and availability is not currently available.