MilliporeSigma
  • Current concepts in systemic and topical therapy for superficial mycoses.
Current concepts in systemic and topical therapy for superficial mycoses.
Clinics in dermatology (2010-03-30)
Larry E Millikan
ABSTRACT
There presently exists a wide selection of choices in the treatment of superficial mycoses. The main categories of broad-spectrum agents are the allylamines and imidazoles, which have been tried and proven over more than 2 decades of usage with good safety. Nystatin and griseofulvin have even longer experience of about 5 decades but have niche usage for yeasts and dermatophytes, respectively. Although no new therapeutic groups have appeared, extensive development of vehicles and delivery systems has enhanced therapeutic results and increased patient compliance.