MilliporeSigma
  • Langmuir-Blodgett films of pyridyldithio-modified multiwalled carbon nanotubes as a support to immobilize hydrogenase.
Langmuir-Blodgett films of pyridyldithio-modified multiwalled carbon nanotubes as a support to immobilize hydrogenase.
Langmuir : the ACS journal of surfaces and colloids (2010-04-02)
Qing Sun, Nikolay A Zorin, Dan Chen, Meng Chen, Tian-Xi Liu, Jun Miyake, Dong-Jin Qian
ABSTRACT
Pyridylthio-modified multiwalled carbon nanotubes (pythio-MWNTs) have been prepared by a reaction of the oxidized MWNTs with S-(2-aminoethylthio)-2-thiopyridine hydrochloride. The obtained pythio-MWNTs nanocomposites formed stable floating monolayers at the air-water interface, which were transferred onto substrate surfaces by the Langmuir-Blodgett (LB) method. Compositions and morphologies of the LB films were characterized by absorption, Raman, X-ray photoelectron spectra as well as by scan electron microscopy and atomic force microscopy. These pythio-MWNTs LB films were then used as a support to immobilize hydrogenase (H(2)ase) to form bionanocomposite of pythio-MWNTs-H(2)ase. Cyclic voltammograms for indium tin oxide electrode covered with the pythio-MWNTs-H(2)ase films were investigated in both Ar and H(2) saturated 0.05 M KCl electrolyte solutions at pH from 4.0 to 9.0. A reversible redox couple of [4Fe-4S](2+/1+) clusters of H(2)ase was recorded when the pH value was 6.0 and 9.0, with reduction and oxidation potentials appearing at about -0.70 and -0.35 V vs Ag/AgCl, respectively. It was revealed that the H(2)ase was of high catalytic activity and strong stability in the LB films of pythio-MWNTs-H(2)ase. Hence, we suggested that the present bionanocomposites could be used as heterogeneous biocatalyst to catalyze reversible reaction between protons and H(2), resulting in potential applications in biohydrogen evolution and H(2) biofuel cells.
Sigma-Aldrich
2-Mercaptopyridine, ReagentPlus®, 99%