MilliporeSigma
  • Dimeric quinidine-catalyzed enantioselective aminooxygenation of oxindoles: an organocatalytic approach to 3-hydroxyoxindole derivatives.
Dimeric quinidine-catalyzed enantioselective aminooxygenation of oxindoles: an organocatalytic approach to 3-hydroxyoxindole derivatives.
Journal of the American Chemical Society (2010-04-02)
Tommy Bui, Nuno R Candeias, Carlos F Barbas
ABSTRACT
3-Hydroxyoxindoles are common structural motifs found in a vast array of natural and biologically active molecules. Most catalytic methods for the asymmetric syntheses of these compounds require the use of transition-metal catalysts. In contrast, alternative catalytic procedures involving organocatalysis are scarce. Herein we disclose a novel aminooxygenation of oxindoles with nitrosobenzene catalyzed by a newly designed quinidine dimer to afford the desired products in good yields with enantioselectivities up to 96%. These reactions allow one to construct a C-O bond at the C(3) position of oxindoles with the creation of an oxygen-containing tetrasubstituted chiral center and provide a new, general organocatalytic approach to the synthesis of 3-hydroxyoxindole derivatives.
Sigma-Aldrich
Nitrosobenzene, ≥97%