MilliporeSigma
  • A novel sensor for cephalosporins based on electrocatalytic oxidation by poly(o-anisidine)/SDS/Ni modified carbon paste electrode.
A novel sensor for cephalosporins based on electrocatalytic oxidation by poly(o-anisidine)/SDS/Ni modified carbon paste electrode.
Talanta (2010-05-06)
Reza Ojani, Jahan-Bakhsh Raoof, Saeed Zamani
ABSTRACT
In this work for first time, the electrocatalytic oxidations of some cephalosporins were carried out by poly(o-anisidine)/SDS/Ni modified carbon paste electrode using cyclic voltammetry, chronoamperometry and chronocoulometry methods. At first, poly(o-anisidine) was formed by cyclic voltammetry in monomer solution containing sodium dodesyl sulfate (SDS), on carbon paste electrode surface. Then, Ni(II) ions were incorporated to electrode by immersion of the polymeric modified electrode having amine group in 0.1molL(-1) Ni(II) ion solution. A good redox behavior was observed for the Ni(OH)(2)/NiOOH couple on the surface of this electrode. Cephalosporins were successfully oxidized on the surface of this nickel ions dispersed poly(o-anisidine) modified carbon paste electrode. The electrocatalytic oxidation peak currents of cephalosporins were linearly dependent on their concentration. Electrode was successfully applied to determine cephalosporins in pharmaceutical preparations.
Sigma-Aldrich
o-Anisidine, ≥99%