MilliporeSigma
  • Amino acyl conjugates of nitrogen heterocycles as potential pharmacophores.
Amino acyl conjugates of nitrogen heterocycles as potential pharmacophores.
The Journal of organic chemistry (2010-05-13)
Alan R Katritzky, Kiran Bajaj, Mael Charpentier, Ebrahim Ghazvini Zadeh
ABSTRACT
2-Methyl- and 4-methylpyridine and 2-methylquinoline are converted by benzotriazole-activated (Cbz)-protected amino acids into chiral potential novel pharmacophore aminoacyl conjugates (33-53%).
Sigma-Aldrich
Quinaldine, ≥95.0% (GC)
Supelco
Quinaldine, analytical standard, ≥90% (GC)