MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • [Effect of amygdalin on serum proteinic biomarker in pulmonary fibrosis of bleomycin-induced rat].
[Effect of amygdalin on serum proteinic biomarker in pulmonary fibrosis of bleomycin-induced rat].
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases (2010-05-15)
Hai-Ke Du, Fu-Cheng Song, Xin Zhou, He Li, Jian-Peng Zhang
ABSTRACT
To evaluate effect of amygdalin on expression of four biomarkers in the animal model of pulmonary fibrosis induced by bleomycin. Rats were given one dose (5 mg/kg) of bleomycin in bleomycin-treated groups, amygdalin-treated groups and saline in controls by intratracheal instillation exposed surgically. The amygdalin-treated groups rats were treated with intraperitoneal injection of amygdalin (15 mg x kg(-1) x day(-1)). The rats were sacrificed 7, 14 and 28 days after bleomycin administration. Polarized light microscopy and Image-Pro Plus detected I and III collagen expressed in Paraffin-embedded lung sections stained with Sirius red. Surface-enhanced laser desorption-ionization time-of-flight mass spectrometry (SELDI-TOF MS) with weak cationic proteinchip (CM10) detected differentially expressed proteins in the pooled serum samples of all groups. Consistent fibrotic responses were found in all bleomycin and amygdalin-tread groups. On the 7th, 14th and 28th day after bleomycin or saline instillation, four differentially expressed proteins were detected in the pooled serum of all groups rats, consisting of 4 proteins with mass/charge ratio of 3530.7, 7043.5, 8332.6 and 9068.0, respectively. Compared with control groups, protein peaks intensity ratio with mass/charge ratio of 3530.7 on 7, 28 d and 7043.5, 8332.6 and 9068.0 on 7, 14 and 28 d was > 2 in bleomycin-treated groups. Compared with amygdalin-treated groups, protein peaks intensity with mass/charge ratio of 3530.7 at 7, 14, 28 d had no change almost, but protein peaks intensity ratio with mass/charge ratio of 7043.5 at 7 d, 8332.6 on 28 d and 9068.0 on 14 d was > 2 in bleomycin-tread groups. All the four protein peaks intensity had no change almost at other point. Amygdalin may reduce the bleomycin-induced increase of differentially expressed protein peak intensities in rat serum.
Sigma-Aldrich
Amygdalin, BioXtra, ≥97.0% (HPLC)
Sigma-Aldrich
Amygdalin, ≥99% (HPLC), from apricot kernels