MilliporeSigma
  • Involvement of monoaminergic system in the antidepressant-like effect of the flavonoid naringenin in mice.
Involvement of monoaminergic system in the antidepressant-like effect of the flavonoid naringenin in mice.
Progress in neuro-psychopharmacology & biological psychiatry (2010-07-07)
Li-Tao Yi, Cheng-Fu Li, Xia Zhan, Cun-Cang Cui, Fei Xiao, Li-Pan Zhou, Yong Xie
ABSTRACT
Dietary flavonoids possess a multiplicity of neuroprotective actions in various central nervous pathophysiological conditions including depression. In this study, the neuropharmacological mechanism of the dietary flavonoid naringenin was investigated in the mouse behavioral models of depression. For this purpose, we investigated the influence of pretreatment with the inhibitors of serotonin or noradrenaline synthesis, p-chlorophenylalanine methyl ester or α-methyl-p-tyrosine, respectively in the anti-immobility effect of naringenin. Compared to the control group, naringenin significantly decreased the immobility time after acute treatment in the mouse tail suspension test (10, 20 and 50 mg/kg), but not in the forced swimming test, without producing locomotor alteration in the open-field test. In addition, pretreatment of mice with p-chlorophenylalanine methyl ester (100 mg/kg) or α-methyl-p-tyrosine (100 mg/kg) prevented the anti-immobility effect of naringenin (20 mg/kg) in the tail suspension test. Taken together, this data demonstrated that naringenin possessed potent antidepressant-like property via the central serotonergic and noradrenergic systems. Thus, our findings suggest the therapeutic potential of this dietary flavonoid in central nervous system disorders especially depression where monoaminergic systems are involved.
Sigma-Aldrich
4-Chloro-DL-phenylalanine methyl ester hydrochloride