MilliporeSigma
Acute and sub-chronic toxicity of D-allose in rats.
Bioscience, biotechnology, and biochemistry (2010-07-14)
Yusuke Iga, Kazunori Nakamichi, Yoko Shirai, Tatsuhiro Matsuo
ABSTRACT
We examined the acute and sub-chronic toxicity of D-allose in rats. In the acute toxicity test, the calculated LD(50) value was 20.5 g/kg. In the sub-chronic toxicity test, no difference was found among the four groups in most of the serum chemical and hematological test results. These results suggest that D-allose is not toxic to rats.
Sigma-Aldrich
D-Allose, 98%