MilliporeSigma
  • X-ray structure and mutational analysis of the atrazine Chlorohydrolase TrzN.
X-ray structure and mutational analysis of the atrazine Chlorohydrolase TrzN.
The Journal of biological chemistry (2010-07-28)
Jennifer L Seffernick, Erik Reynolds, Alexander A Fedorov, Elena Fedorov, Steven C Almo, Michael J Sadowsky, Lawrence P Wackett
ABSTRACT
Atrazine chlorohydrolase, TrzN (triazine hydrolase or atrazine chlorohydrolase 2), initiates bacterial metabolism of the herbicide atrazine by hydrolytic displacement of a chlorine substituent from the s-triazine ring. The present study describes crystal structures and reactivity of wild-type and active site mutant TrzN enzymes. The homodimer native enzyme structure, solved to 1.40 Å resolution, is a (βα)(8) barrel, characteristic of members of the amidohydrolase superfamily. TrzN uniquely positions threonine 325 in place of a conserved aspartate that ligates the metal in most mononuclear amidohydrolases superfamily members. The threonine side chain oxygen atom is 3.3 Å from the zinc atom and 2.6 Å from the oxygen atom of zinc-coordinated water. Mutation of the threonine to a serine resulted in a 12-fold decrease in k(cat)/K(m), largely due to k(cat), whereas the T325D and T325E mutants had immeasurable activity. The structure and kinetics of TrzN are reminiscent of carbonic anhydrase, which uses a threonine to assist in positioning water for reaction with carbon dioxide. An isosteric substitution in the active site glutamate, E241Q, showed a large diminution in activity with ametryn, no detectable activity with atratone, and a 10-fold decrease with atrazine, when compared with wild-type TrzN. Activity with the E241Q mutant was nearly constant from pH 6.0 to 10.0, consistent with the loss of a proton-donating group. Structures for TrzN-E241Q were solved with bound ametryn and atratone to 1.93 and 1.64 Å resolution, respectively. Both structure and kinetic determinations suggest that the Glu(241) side chain provides a proton to N-1 of the s-triazine substrate to facilitate nucleophilic displacement at the adjacent C-2.
Product Description
Supelco
Ametryn, PESTANAL®, analytical standard