MilliporeSigma
  • Synthesis of 2,3-dihydroquinazolin-4(1H)-ones by three-component coupling of isatoic anhydride, amines, and aldehydes catalyzed by magnetic Fe(3)O(4) nanoparticles in water.
Synthesis of 2,3-dihydroquinazolin-4(1H)-ones by three-component coupling of isatoic anhydride, amines, and aldehydes catalyzed by magnetic Fe(3)O(4) nanoparticles in water.
Journal of combinatorial chemistry (2010-08-06)
Zhan-Hui Zhang, Hong-Yan Lü, Shu-Hong Yang, Jian-Wu Gao
ABSTRACT
A simple and efficient protocol for one-pot three-component coupling of isatoic anhydride, amines, and aldehydes in water using magnetically recoverable Fe(3)O(4) nanoparticles is reported. This methodology results in the synthesis of a variety of 2,3-dihydroquinazolin-4(1H)-ones in high yields. The catalyst can be recovered and recycled without a significant loss in the catalytic activity.
Sigma-Aldrich
Isatoic anhydride, 96%