MilliporeSigma
Tripodal transmembrane transporters for bicarbonate.
Chemical communications (Cambridge, England) (2010-08-10)
Nathalie Busschaert, Philip A Gale, Cally J E Haynes, Mark E Light, Stephen J Moore, Christine C Tong, Jeffery T Davis, William A Harrell
ABSTRACT
Easy-to-make tripodal tris-thiourea receptors based upon tris(2-aminoethyl)amine are capable of chloride/bicarbonate transport and as such represent a new class of bicarbonate transport agent.
Sigma-Aldrich
Tris(2-aminoethyl)amine, 96%