MilliporeSigma
Total syntheses of tubulysins.
Chemistry (Weinheim an der Bergstrasse, Germany) (2010-08-25)
Taku Shibue, Toshihiro Hirai, Iwao Okamoto, Nobuyoshi Morita, Hyuma Masu, Isao Azumaya, Osamu Tamura
ABSTRACT
The total syntheses of tetrapeptides tubulysins D (1 b), U (1 c), and V (1 d), which are potent tubulin polymerization inhibitors, are described. The synthesis of Tuv (2), an unusual amino acid constituent of tubulysins, includes an 1,3-dipolar cycloaddition reaction of chiral nitrone D-6 derived from D-gulose with N-acryloyl camphor sultam (-)-9 employing the double asymmetric induction, whereas the synthesis of Tup (20), another unusual amino acid, involves a stereoselective Evans aldol reaction of (Z)-boron enolate generated from (S)-4-isopropyl-3-propionyl-2-oxazolidinone with N-protected phenylalaninal and a subsequent Barton deoxygenation protocol. We accomplished the total syntheses of tubulysins U (1 c) and V (1 d) by using these methodologies, in which the isoxazolidine ring was used as the effective protective group for γ-amido alcohol functionality. Furthermore, to understand the structure-activity relationship of tubulysins, we synthesized tubulysin D (1 b) and cyclo-tubulysin D (1 e) from 2-Me and 20, and ent-tubulysin D (ent-1 d) from ent-2-Me and ent-20, respectively. The preliminary results regarding their biological activities are also reported.