MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Substrate binding mechanism of a type I extradiol dioxygenase.
The Journal of biological chemistry (2010-09-03)
Hyo Je Cho, Kyungsun Kim, Seo Yean Sohn, Ha Yeon Cho, Kyung Jin Kim, Myung Hee Kim, Dockyu Kim, Eungbin Kim, Beom Sik Kang
ABSTRACT
A meta-cleavage pathway for the aerobic degradation of aromatic hydrocarbons is catalyzed by extradiol dioxygenases via a two-step mechanism: catechol substrate binding and dioxygen incorporation. The binding of substrate triggers the release of water, thereby opening a coordination site for molecular oxygen. The crystal structures of AkbC, a type I extradiol dioxygenase, and the enzyme substrate (3-methylcatechol) complex revealed the substrate binding process of extradiol dioxygenase. AkbC is composed of an N-domain and an active C-domain, which contains iron coordinated by a 2-His-1-carboxylate facial triad motif. The C-domain includes a β-hairpin structure and a C-terminal tail. In substrate-bound AkbC, 3-methylcatechol interacts with the iron via a single hydroxyl group, which represents an intermediate stage in the substrate binding process. Structure-based mutagenesis revealed that the C-terminal tail and β-hairpin form part of the substrate binding pocket that is responsible for substrate specificity by blocking substrate entry. Once a substrate enters the active site, these structural elements also play a role in the correct positioning of the substrate. Based on the results presented here, a putative substrate binding mechanism is proposed.
Sigma-Aldrich
3-Methylcatechol, 98%