MilliporeSigma
  • A HPLC-UV method for the determination of puerarin in rat plasma after intravenous administration of PEGylated puerarin conjugate.
A HPLC-UV method for the determination of puerarin in rat plasma after intravenous administration of PEGylated puerarin conjugate.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-11-09)
Xinyi Liu, Hongying Zhi, Feng Du, Zuguang Ye, Naijie Wang, Wenjie Qin, Jianrong Li
ABSTRACT
A sensitive and reproducible HPLC method for quantitative determination of puerarin (PUE) in rat plasma was developed and validated using 4-hydroxybenzaldehyde as an internal standard. The separation of PUE was performed on a CAPCELL PAK C18 column by gradient elution with 0.2% aqueous phosphoric acid and acetonitrile as the mobile phase. The method was validated and found to be linear in the range of 80-12,000ng/mL. The limit of quantification was 80ng/mL based on 100μL of plasma. The variations for intra- and inter-day precision were less than 8.3%, and the accuracy values were between 98% and 105.2%. The extraction recoveries were more than 85%. The method was successfully applied in the comparative study of pharmacokinetics of PEGylated puerarin (PEG-PUE) versus PUE in rats. Compared with PUE, PEG-PUE showed a 5.2-fold increase in half-life of PUE and a 4.7-fold increase in mean residence time. In addition, this method was also successfully applied to determine the low plasma concentration of PUE regenerated from PEG-PUE in vitro.
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥95.0% (HPLC)
Sigma-Aldrich
4-Hydroxybenzaldehyde, 98%
Sigma-Aldrich
4-Hydroxybenzaldehyde, ≥97%, FG