MilliporeSigma
  • Degradation of dichloroaniline isomers by a newly isolated strain, Bacillus megaterium IMT21.
Degradation of dichloroaniline isomers by a newly isolated strain, Bacillus megaterium IMT21.
Microbiology (Reading, England) (2010-12-18)
Xie-Feng Yao, Fazlurrahman Khan, Rinku Pandey, Janmejay Pandey, Roslyn G Mourant, Rakesh K Jain, Jian-Hua Guo, Robyn J Russell, John G Oakeshott, Gunjan Pandey
ABSTRACT
An efficient 3,4-dichloroaniline (3,4-DCA)-mineralizing bacterium has been isolated from enrichment cultures originating from a soil sample with a history of repeated exposure to diuron, a major metabolite of which is 3,4-DCA. This bacterium, Bacillus megaterium IMT21, also mineralized 2,3-, 2,4-, 2,5- and 3,5-DCA as sole sources of carbon and energy. These five DCA isomers were degraded via two different routes. 2,3-, 2,4- and 2,5-DCA were degraded via previously unknown dichloroaminophenol metabolites, whereas 3,4- and 3,5-DCA were degraded via dichloroacetanilide.
Sigma-Aldrich
3,4-Dichloroaniline, 98%