MilliporeSigma
  • A validated enantioselective assay for the simultaneous quantitation of (R)-, (S)-fluoxetine and (R)-, (S)-norfluoxetine in ovine plasma using liquid chromatography with tandem mass spectrometry (LC/MS/MS).
A validated enantioselective assay for the simultaneous quantitation of (R)-, (S)-fluoxetine and (R)-, (S)-norfluoxetine in ovine plasma using liquid chromatography with tandem mass spectrometry (LC/MS/MS).
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-01-19)
Timothy W Chow, András Szeitz, Dan W Rurak, K Wayne Riggs
ABSTRACT
A liquid chromatography-tandem mass spectrometry (LC/MS/MS) method was developed and validated for the quantitation of (R)-, (S)-fluoxetine, and (R)-, (S)-norfluoxetine in ovine plasma. The analytes were extracted from ovine plasma at a basic pH using a single-step liquid-liquid extraction with methyl-tert-butyl ether. Chromatographic separation of all enantiomers was achieved using an AGP-chiral column with a run time of 10 min. (R)-, (S)-fluoxetine, and (R)-, (S)-norfluoxetine were quantitated at the total ion current (TIC) of multiple reaction monitoring (MRM) transitions of m/z 310.2→44.1, m/z 310.2→147.7 for (R)-, (S)-fluoxetine, and m/z 296.2→30.3, m/z 296.2→133.9 for (R)-, (S)-norfluoxetine. This method was validated for accuracy, precision, linearity, range, limit of quantitation (LOQ), selectivity, recovery, dilution integrity, matrix effect, and evaluation of carry-over. Observed accuracy ranges were as follows: (R)-fluoxetine -8.82 to 3.75%; (S)-fluoxetine -10.8 to 1.46%; (R)-norfluoxetine -7.50 to 0.37% and (S)-norfluoxetine -8.77% to -1.33%. Observed precision ranges were as follows: (R)-fluoxetine 5.29-11.5%; (S)-fluoxetine 3.91-11.1%; (R)-norfluoxetine 4.32-7.67% and (S)-norfluoxetine -8.77% to -1.33%. The calibration curves were weighted (1/X(2), n=4) and observed to be linear for all analytes with the following r(2) values: (R)-fluoxetine ≥ 0.997; (S)-fluoxetine ≥ 0.996; (R)-norfluoxetine ≥ 0.989 and (S)-norfluoxetine ≥ 0.994. The analytical range of the method was 1-500 ng/ml with an LOQ of 1 ng/ml for all analytes, using a sample volume of 300 μL.