MilliporeSigma
  • Coupling to polymeric scaffolds stabilizes biofunctional peptides for intracellular applications.
Coupling to polymeric scaffolds stabilizes biofunctional peptides for intracellular applications.
Molecular pharmacology (2011-01-21)
Ivo R Ruttekolk, Alokta Chakrabarti, Martin Richter, Falk Duchardt, Heike Glauner, Wouter P R Verdurmen, Jörg Rademann, Roland Brock
ABSTRACT
Here, we demonstrate that coupling to N-hydroxypropyl methacrylamide (HPMA) copolymer greatly enhances the activity of apoptosis-inducing peptides inside cells. Peptides corresponding to the BH3 domain of Bid were coupled to a thioester-activated HPMA (28.5 kDa) via native chemical ligation in a simple one-pot synthesis. Peptides and polymer conjugates were introduced into cells either by electroporation or by conjugation to the cell-penetrating peptide nona-arginine. The molecular basis of the increased activity is elucidated in detail. Loading efficiency and intracellular residence time were assessed by confocal microscopy. Fluorescence correlation spectroscopy was used as a separation-free analytical technique to determine proteolytic degradation in crude cell lysates. HPMA conjugation strongly increased the half-life of the peptides in crude cell lysates and inside cells, revealing proteolytic protection as the basis for higher activity.
Sigma-Aldrich
Hydroxypropyl methacrylate, Mixture of hydroxypropyl and hydroxyisopropyl methacrylates, 97%, contains 180-220 ppm monomethyl ether hydroquinone as inhibitor