MilliporeSigma
  • Efficacy of allicin from garlic against Ascaridia galli infection in chickens.
Efficacy of allicin from garlic against Ascaridia galli infection in chickens.
Poultry science (2011-01-21)
F C Velkers, K Dieho, F W M Pecher, J C M Vernooij, J H H van Eck, W J M Landman
ABSTRACT
The use of garlic as a treatment against helminth infections is increasing in organic layer farms in several European countries. Its efficacy against these parasites, however, has not been demonstrated thus far. Therefore, a study was conducted to determine the efficacy of a commercially available garlic product consisting of a high concentration of allicin (i.e., the main active component of garlic) against experimentally induced Ascaridia galli infection in chickens. In total, 450 Lohmann LSL-Classic cockerels were used. Group 1, the uninfected, untreated group, consisted of 50 chickens. Groups 2 to 5, each consisting of approximately 100 chickens, were inoculated with 300 embryonated A. galli eggs/chicken at 6 wk of age. Group 2 was not treated, whereas groups 3 through 5 were given daily individual oral treatments from 13 wk of age onward. Group 3 received the recommended dose of allicin for 2 wk, whereas group 4 received a 10-fold dose of allicin. Group 5 was given 10 mg of flubendazole/kg of BW for 1 wk. Necropsy of 20 birds of all groups was performed weekly between 13 and 16 wk of age to determine adult worm loads. Group 1 remained free of A. galli. The experimental infection in the other groups resulted in a mean adult worm load of approximately 16 worms/bird. No significant differences were observed in worm counts of the allicin-treated groups (groups 3 and 4) compared with the infected, untreated group (group 2) at any week (P > 0.05). In contrast, no worms were found in chickens after flubendazole treatment (group 5). It was concluded that allicin does not represent an alternative to flubendazole for the treatment of A. galli infections in chickens.
Product Description
Supelco
Flubendazol, VETRANAL®, analytical standard