MilliporeSigma
  • Synthesis and biological evaluation of novel acenaphthene derivatives as potential antitumor agents.
Synthesis and biological evaluation of novel acenaphthene derivatives as potential antitumor agents.
Molecules (Basel, Switzerland) (2011-03-19)
Yong-Mei Xie, Yi Deng, Xiao-Yun Dai, Jie Liu, Liang Ouyang, Yu-Quan Wei, Ying-Lan Zhao
ABSTRACT
Twelve novel acenaphthene derivatives have been synthesized. The structures of all compounds were confirmed by ¹H-NMR, MS and elemental analysis. Their antitumor activities were evaluated in six human solid tumor cell lines, namely non-small cell lung cancer (H460), human colon adenocarcinoma (SW480), human breast cancer cell (MDA-MB-468 and SKRB-3), human melanoma cell (A375) and human pancreatic cancer (BxPC-3). Among them, compound 3c shows the best antitumor activity against SKRB-3 cell line, as high as the positive control adriamycin.
Sigma-Aldrich
Acenaphthene, 99%
Supelco
Acenaphthene, analytical standard
Supelco
Acenaphthene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland