MilliporeSigma
  • HPMA based amphiphilic copolymers mediate central nervous effects of domperidone.
HPMA based amphiphilic copolymers mediate central nervous effects of domperidone.
Macromolecular rapid communications (2011-04-07)
Mirjam Hemmelmann, Christiane Knoth, Ulrich Schmitt, Mareli Allmeroth, Dorothea Moderegger, Matthias Barz, Kaloian Koynov, Christoph Hiemke, Frank Rösch, Rudolf Zentel
ABSTRACT
In this study we give evidence that domperidone encapsulated into amphiphilic p(HPMA)-co-p(laurylmethacrylate) (LMA) copolymer aggregates is able to cross the blood-brain barrier, since it affected motor behaviour in animals, which is a sensitive measure for CNS actions. Carefully designed copolymers based on the clinically approved p(HPMA) were selected and synthesized by a combination of controlled radical polymerization and post-polymerization modification. The hydrodynamic radii (R(h) ) of amphiphilic p(HPMA)-co-p(LMA) alone and loaded with domperidone were determined by fluorescence correlation spectroscopy.
Sigma-Aldrich
Hydroxypropyl methacrylate, Mixture of hydroxypropyl and hydroxyisopropyl methacrylates, 97%, contains 180-220 ppm monomethyl ether hydroquinone as inhibitor