MilliporeSigma
  • Fungi as a promising tool for bioremediation of soils contaminated with aromatic amines, a major class of pollutants.
Fungi as a promising tool for bioremediation of soils contaminated with aromatic amines, a major class of pollutants.
Nature reviews. Microbiology (2011-04-19)
Philippe Silar, Julien Dairou, Angélique Cocaign, Florent Busi, Fernando Rodrigues-Lima, Jean-Marie Dupret
Sigma-Aldrich
3,4-Dichloroaniline, 98%