MilliporeSigma
  • Possible factors influencing the separation mechanism of diastereomeric amino acid derivatives obtained from s-triazine type reagents.
Possible factors influencing the separation mechanism of diastereomeric amino acid derivatives obtained from s-triazine type reagents.
Journal of chromatography. A (2011-05-03)
Eric Loeser
ABSTRACT
Diastereomeric derivatives prepared from an amino acid and an amino amide using trichloro s-triazine as a coupling platform are known to produce good chromatographic diastereoselectivity for many amino acid analytes. The chromatographic diastereoselectivity of these derivatives is difficult to rationalize based on the possibility of four possible conformational isomers, which can interconvert by rotation about the C-N bonds between the amino substituents and the triazine ring. The observed diastereoselectivity implicates an unobvious but significant driving force which causes one of several conformations to be favored over the others. Several possibilities are discussed. Intramolecular hydrogen bonding between acid and amide substituents was explored using computer aided molecular modeling. While such hydrogen bonding may be geometrically possible between the amino acid and the amide substituents, it does not explain why derivatives produced from other chiral compounds which are not capable of the same hydrogen bonding interaction nevertheless exhibit substantial diastereoselectivity. Two other more general effects, steric hindrance to solvation and ion pairing, are therefore suggested as possible contributing factors to the chromatographic diastereoselectivity. Based on the conformational equilibrium behavior of related triazine compounds as reported in the literature, either one of these effects could influence the conformation of the diastereomeric derivatives even in the absence of intramolecular hydrogen bonding interactions between the two chiral substituents, and these effects may therefore be a contributing factor for the observed elution order of the diastereomers.
Sigma-Aldrich
Cyanuric chloride, 99%