MilliporeSigma
  • Targeted synthesis of a 2D ordered porous organic framework for drug release.
Targeted synthesis of a 2D ordered porous organic framework for drug release.
Chemical communications (Cambridge, England) (2011-05-10)
Huanyu Zhao, Zhao Jin, Hongmin Su, Xiaofei Jing, Fuxing Sun, Guangshan Zhu
ABSTRACT
A novel 2D porous organic framework based on the nucleophilic substitution of cyanuric chloride has been designed and synthesized successfully, which possesses an ordered structure, permanent porosity and drug release ability towards ibuprofen.
Sigma-Aldrich
Cyanuric chloride, 99%