MilliporeSigma
  • A transparent photo-responsive organogel based on a glycoluril supergelator.
A transparent photo-responsive organogel based on a glycoluril supergelator.
Chemical communications (Cambridge, England) (2011-06-02)
Konrad Tiefenbacher, Henry Dube, Dariush Ajami, Julius Rebek
ABSTRACT
The synthesis of an azo-benzene glycoluril supergelator is reported. The control over the gel/sol state can be accomplished by irradiation and heat, but also (in a unidirectional sense) by incorporation of the glycoluril into a more stable supramolecular assembly.
Sigma-Aldrich
Glycoluril, 97%