MilliporeSigma
  • Three phases hollow fiber LPME combined with HPLC-UV for extraction, preconcentration and determination of valerenic acid in Valeriana officinalis.
Three phases hollow fiber LPME combined with HPLC-UV for extraction, preconcentration and determination of valerenic acid in Valeriana officinalis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2011-06-07)
Mohamad Mirzaei, Hossein Dinpanah
ABSTRACT
In the present work, the applicability of hollow fiber-based liquid phase microextraction (HF-LPME) was evaluated for the extraction and preconcentration of valerenic acid prior to its determination by reversed-phase HPLC/UV. The target drug was extracted from 5.0 mL of aqueous solution with pH 3.5 into an organic extracting solvent (dihexyl ether) impregnated in the pores of a hollow fiber and finally back extracted into 10 μ L of aqueous solution with pH 9.5 located inside the lumen of the hollow fiber. In order to obtain high extraction efficiency, the parameters affecting the HF-LPME, including pH of the donor and acceptor phases, type of organic phase, ionic strength, the volume ratio of donor to acceptor phase, stirring rate and extraction time were studied and optimized. Under the optimized conditions, enrichment factor up to 446 was achieved and the relative standard deviation (RSD) of the method was 4.36% (n = 9). The linear range was 7.5-850 μg L⁻¹ with correlation coefficient (r²=0.999), detection limits was 2.5 μg L⁻¹ and the LOQ was 7.5 μg L⁻¹. The proposed method was evaluated by extraction and determination of valerenic acid in some Iranian wild species of Valerianaceae.
Product Description
Valerenic acid, primary reference standard
Supelco
Valerenic acid, analytical standard