MilliporeSigma
  • Role of reactive oxygen and nitrogen species in olfactory epithelial injury by the sulfur mustard analogue 2-chloroethyl ethyl sulfide.
Role of reactive oxygen and nitrogen species in olfactory epithelial injury by the sulfur mustard analogue 2-chloroethyl ethyl sulfide.
American journal of respiratory cell and molecular biology (2011-06-07)
Heidi C O'Neill, David J Orlicky, Tara B Hendry-Hofer, Joan E Loader, Brian J Day, Carl W White
ABSTRACT
The inhalation of sulfur mustard (SM) causes substantial deposition in the nasal region. However, specific injury has not been characterized. 2-chloroethyl ethyl sulfide (CEES) is an SM analogue used to model injury and screen potential therapeutics. After the inhalation of CEES, damage to the olfactory epithelium (OE) was extensive. Terminal deoxynucleotidyl transferase-mediated dUTP nick-end labeling-positive cells were present by 4 hours, and maximal at 18-72 hours. Cleaved caspase 3 immunohistochemistry (IHC) was maximal at 18 hours after the inhalation of 5% CEES. Olfactory marker protein (OMP)-positive olfactory neurons were markedly decreased at 18 hours. IHC-positive cells for 3-nitrotyrosine (3-NT) within epithelium were elevated by 8 hours, waning by 18 hours, and absent by 72 hours. AEOL 10150, a catalytic manganoporphyrin antioxidant, administered both subcutaneously (5 mg/kg) and intranasally (50 μM, "combined treatment"), decreased OE injury. CEES-induced increases in markers of cell death were decreased by combined treatment involving AEOL 10150. CEES-induced changes in OMP and 3-NT immunostaining were markedly improved by combined treatment involving AEOL 10150. The selective inducible nitric oxide synthase inhibitor 1400W (5 mg/kg, subcutaneous), administered 1 hour after inhalation and thereafter every 4 hours (five doses), also reduced OE damage with improved OMP and 3-NT staining. Taken together, these data indicate that reactive oxygen and nitrogen species are important mediators in CEES-induced nasal injury.
Sigma-Aldrich
2-Chloroethyl ethyl sulfide, 97%