MilliporeSigma
  • Synthesis of mesoporous β-Ga2O3 nanorods using PEG as template: preparation, characterization and photocatalytic properties.
Synthesis of mesoporous β-Ga2O3 nanorods using PEG as template: preparation, characterization and photocatalytic properties.
Journal of hazardous materials (2011-07-20)
Weirong Zhao, Yong Yang, Rui Hao, Feifei Liu, Yan Wang, Min Tan, Jing Tang, Daqing Ren, Dongye Zhao
ABSTRACT
Mesoporous wide bandgap semiconductors offer high photocatalytic oxidation and mineralization activities. In this study, mesoporous β-Ga(2)O(3) diamond nanorods with 200-300 nm in diameter and 1.0-1.2 μm in length were synthesized via a urea-based hydrothermal method using polyethylene glycol (PEG) as template agent. The UV photocatalytic oxidation activity of β-Ga(2)O(3) for gaseous toluene was evaluated, and 7 kinds of intermediates were monitored online by a proton transfer reaction mass spectrometry. Photoluminescence spectra manifested that the dosage and molecular weight of PEG are crucial for formation of vacancies and photocatalytic oxidation activities. A PEG-assisted hydrothermal formation mechanism of mesoporous β-Ga(2)O(3) diamond nanorods was proposed. Based on the health risk influence index (η) of the intermediates, the calculated health risks revealed that the β-Ga(2)O(3) nanorods with a η value of 9.6 are much safer than TiO(2) (η = 17.6).
Sigma-Aldrich
Gallium(III) oxide, ≥99.99% trace metals basis