MilliporeSigma
  • Synthesis and characterization of a Eu-containing polymer precursor featuring thenoyltrifluoroacetone and 5-acrylamido-1,10-phenanthroline.
Synthesis and characterization of a Eu-containing polymer precursor featuring thenoyltrifluoroacetone and 5-acrylamido-1,10-phenanthroline.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-08-02)
Cun-Jin Xu, Bo-Geng Li, Jin-Tao Wan, Zhi-Yang Bu
ABSTRACT
A polymerizable ligand, 5-acrylamido-1,10-phenanthroline (L), was synthesized. Its Eu(III) complex with 2-thenoyltrifluoroacetone (HTTA) was prepared and characterized by elemental analysis, IR, MS, and (1)H NMR spectra. The photophysical properties of the complex were studied in detail by using UV, luminescence spectra, luminescence lifetime and quantum yield. The complex shows a remarkable luminescence quantum yield at room temperature (40.1%) upon ligand excitation and a long (5)D(0) lifetime (590 μs), which makes it not only a promising light-conversion molecular device but also an excellent luminescent polymer precursor.
Sigma-Aldrich
2-Thenoyltrifluoroacetone, 99%