MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Oxidation of 2,6-dimethylaniline by the Fenton, electro-Fenton and photoelectro-Fenton processes.
Oxidation of 2,6-dimethylaniline by the Fenton, electro-Fenton and photoelectro-Fenton processes.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2011-08-03)
Wang-Ping Ting, Yao-Hui Huang, Ming-Chun Lu
ABSTRACT
Fenton technologies for wastewater treatment have demonstrated their effectiveness in eliminating toxic compounds. This study examines how hydrogen peroxide concentration and ultraviolet (UV) light affects oxidation processes. However, total mineralization through these Fenton technologies is expensive compared with biological technologies. Therefore, partial chemical oxidation of toxic wastewaters with Fenton processes followed by biological units may increase the application range of Fenton technologies. Using 2,6-dimethylaniline (2,6-DMA) as the target compound, this study also investigates oxidation intermediates and their biodegradable efficiencies after treatment by Fenton, electro-Fenton and photoelectron-Fenton processes. Analytical results show that the UV light-promoting efficiency, r(PE-F)/r(E-F), was 2.02, 2.55 and 2.67 with initial hydrogen peroxide concentrations of 15, 20 and 25 mM, respectively. We conclude that UV irradiation promoted 2,6-DMA degradation significantly. The same tendency was observed for biochemical oxygen demand/total organic carbon (BOD(5)/TOC) ratios for each process, meaning that 2,6-DMA can be successfully detoxified using the electro-Fenton and photoelectro-Fenton processes. Some organic intermediates aminobenzene, nitrobenzene, 2,6-dimethylphenol, phenol and oxalic acid--were detected in different oxidation processes.
Product Description
Supelco
2,6-Dimethylaniline, PESTANAL®, analytical standard
Sigma-Aldrich
2,6-Dimethylaniline, 99%
Supelco
2,6-Dimethylaniline, analytical standard