MilliporeSigma
  • Determination of dicofol in aquatic products using molecularly imprinted solid-phase extraction coupled with GC-ECD detection.
Determination of dicofol in aquatic products using molecularly imprinted solid-phase extraction coupled with GC-ECD detection.
Talanta (2011-08-30)
Hui Wang, Hongyuan Yan, Mande Qiu, Jindong Qiao, Gengliang Yang
ABSTRACT
The novel molecularly imprinted microsphere (MIM) that could be applied as special sorbent was synthesized by aqueous suspension polymerization using 1,1-bis(4-chlorophenyl)-1,2,2,2-tetrachloroethane (α-chloro-DDT) as the dummy template. The obtained MIM exhibited good recognition and selectivity to dicofol and it was successfully applied as selective sorbent of solid-phase extraction for the determination of dicofol from aquatic products. At the optimum conditions of the molecularly imprinted solid-phase extraction (MISPE) coupled with GC-ECD, good linearity for dicofol was achieved in a range of 0.4-100 ng g(-1) (r(2)=0.9995) and the recoveries at three spiked levels were ranged from 85.8% to 101.2% for aquatic products with the relative standard deviation (RSD) less than 5.6%. The presented MISPE-GC-ECD method exhibited the advantages of simplicity, selectivity and sensitivity, and could be potentially applied to the determination of dicofol in complicated aquatic products.
Product Description
Supelco
Dicofol, PESTANAL®