MilliporeSigma
Kinetic cooperativity of tyrosinase. A general mechanism.
Acta biochimica Polonica (2011-09-03)
Jose Luis Muñoz-Muñoz, Francisco Garcia-Molina, Ramón Varon, Jose Tudela, Francisco Garcia-Cánovas, Jose N Rodríguez-López
ABSTRACT
Tyrosinase shows kinetic cooperativity in its action on o-diphenols, but not when it acts on monophenols, confirming that the slow step is the hydroxylation of monophenols to o-diphenols. This model can be generalised to a wide range of substrates; for example, type S(A) substrates, which give rise to a stable product as the o-quinone evolves by means of a first or pseudo first order reaction (α-methyl dopa, dopa methyl ester, dopamine, 3,4-dihydroxyphenylpropionic acid, 3,4-dihydroxyphenylacetic acid, α-methyl-tyrosine, tyrosine methyl ester, tyramine, 4-hydroxyphenylpropionic acid and 4-hydroxyphenylacetic acid), type S(B) substrates, which include those whose o-quinone evolves with no clear stoichiometry (catechol, 4-methylcatechol, phenol and p-cresol) and, lastly, type S(C) substrates, which give rise to stable o-quinones (4-tert-butylcatechol/4-tert-butylphenol).
Sigma-Aldrich
4-Methylcatechol, ≥95%
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
3-(4-Hydroxyphenyl)propionic acid, 98%