MilliporeSigma
  • Interactions of an asymmetric amine with a non-C2 symmetric Cu-salen complex: an EPR/ENDOR and HYSCORE investigation.
Interactions of an asymmetric amine with a non-C2 symmetric Cu-salen complex: an EPR/ENDOR and HYSCORE investigation.
Physical chemistry chemical physics : PCCP (2011-10-14)
Ignacio Caretti, Emma Carter, Ian A Fallis, Damien M Murphy, Sabine Van Doorslaer
ABSTRACT
Single enantiomers of R-/S-methylbenzylamine (MBA) were found to selectively form adducts with the chiral non-C(2) symmetric Cu-salen complex N-(3,5-di-tert-butylsalicylidene)-N'-(salicylidene)-cyclohexane-1,2-diamine copper(II), hereafter labelled [Cu(3)]. The g/A spin Hamiltonian parameters of this Cu(II) complex showed a decrease in symmetry from axial to rhombic upon formation of the [Cu(3)] + MBA adducts. The selectivity in enantiomeric discrimination was found to be only 59 ± 5% in favour of the heterochiral R,R'-[Cu(3)] + S-MBA and S,S'-[Cu(3)] + R-MBA adducts. This was directly evidenced by W-band EPR spectroscopy. The observed low selectivity for enantiomer discrimination is primarily attributed to the loss of the bulky tert-butyl groups from the 3,5 positions of [Cu(3)] compared to the parent N,N'-bis(3,5-di-tert-butylsalicylidene)-cyclohexane-1,2-diamine copper(II) ligand (labelled [Cu(1)]). The structure of the [Cu(3)] complex in the presence and absence of coordinating amine was further investigated by analysis of the ligand hyperfine interactions, as revealed through Q-band CW-ENDOR, X-band Davies ENDOR and HYSCORE. (1)H couplings from the -NH(2) group of the amine, observed by ENDOR and HYSCORE, provided direct evidence of amine coordination.
Sigma-Aldrich
(S)-(−)-α-Methylbenzylamine, 98%
Sigma-Aldrich
1,2-Diaminocyclohexane, mixture of cis and trans, 99%
Sigma-Aldrich
(1S,2S)-(+)-1,2-Diaminocyclohexane, 98%
Sigma-Aldrich
α-Methylbenzylamine, 99%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, ChiPros®, produced by BASF, ≥99.0%
Sigma-Aldrich
(1R,2R)-(−)-1,2-Diaminocyclohexane, 98%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, 98%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, purum, ≥98.0% (sum of enantiomers, GC)
Supelco
(R)-(+)-α-Methylbenzylamine, derivatization grade (chiral), LiChropur, ≥99.0%
Sigma-Aldrich
(±)-trans-1,2-Diaminocyclohexane, 99%