MilliporeSigma
  • Simultaneous injection effective mixing analysis system for the determination of direct bilirubin in urinary samples.
Simultaneous injection effective mixing analysis system for the determination of direct bilirubin in urinary samples.
Talanta (2011-11-22)
Kraingkrai Ponhong, Norio Teshima, Kate Grudpan, Shoji Motomizu, Tadao Sakai
ABSTRACT
A novel simultaneous injection effective mixing analysis system (SIEMA) for determination of direct bilirubin in urine sample was developed. Bilirubin reacts with diazotized sulfanilic acid in the presence of n-octyl-β-d-thioglucoside (OTG) as a solubilizing agent to form OTG-azobilirubin. The flow and chemical variables were investigated. A linear calibration graph for direct bilirubin was obtained over the range of 0-1.0 mg L(-1) (r(2)=0.994) with the limit of detection (3σ) of 4.7 μg L(-1), and the relative standard deviation (RSD) being 1.9% (n=11, 0.5 mg L(-1) of direct bilirubin). The results in healthy adult urine obtained by the proposed approach were found in good agreement with those obtained by the batch-wise diazo method.
Sigma-Aldrich
Octyl β-D-1-thioglucopyranoside, ≥98.0% (GC)
Pricing and availability is not currently available.