MilliporeSigma
  • Acute phase protein response in the capybara (Hydrochoerus hydrochaeris).
Acute phase protein response in the capybara (Hydrochoerus hydrochaeris).
Journal of wildlife diseases (2011-11-22)
Luis Bernal, Mariane Feser, Silvia Martínez-Subiela, Juan D García-Martínez, José J Cerón, Fernando Tecles
ABSTRACT
We evaluated the acute phase protein response in capybaras (Hydrochoerus hydrochaeris). Three animal groups were used: 1) healthy animals (n=30), 2) a group in which experimental inflammation with turpentine was induced (n=6), and 3) a group affected with sarcoptic scabies (n=14) in which 10 animals were treated with ivermectin. Haptoglobin (Hp), acid-soluble glycoprotein (ASG) and albumin were analyzed in all animals. In those treated with turpentine, Hp reached its maximum value at 2 wk with a 2.7-fold increase, whereas ASG increased 1.75-fold and albumin decreased 0.87-fold 1 wk after the induction of inflammation. Capybaras affected with sarcoptic scabies presented increases in Hp and ASG of 4.98- and 3.18-fold, respectively, and a 0.87-fold decrease in albumin, compared with healthy animals. Haptoglobin and ASG can be considered as moderate, positive acute phase proteins in capybaras because they showed less than 10-fold increases after an inflammatory process and reached their peak concentrations 1 wk after the induction of inflammation. Conversely, albumin can be considered a negative acute phase protein in capybaras because it showed a reduction in concentration after inflammatory stimulus.
Sigma-Aldrich
Oil of turpentine, SAJ first grade
Sigma-Aldrich
Oil of turpentine, purified