MilliporeSigma
Bioresponsive systems based on polygalacturonate containing hydrogels.
Enzyme and microbial technology (2011-11-25)
Konstantin P Schneider, Alexandra Rollett, Eva Wehrschuetz-Sigl, Andrea Hasmann, Armin Zankel, Andreas Muehlebach, Franz Kaufmann, Georg M Guebitz
ABSTRACT
Polysaccharide acid (PSA) based devices (consisting of alginic acid and polygalacturonic acid) were investigated for the detection of contaminating microorganisms. PSA-CaCl(2) hydrogel systems were compared to systems involving covalent cross-linking of PSA with glycidylmethacrylate (PSA-GMA) which was confirmed with Fourier Transformed Infrared (FTIR) analysis. Incubation of PSA-CaCl(2) and PSA-GMA beads loaded with Alizarin as a model ingredient with trigger enzymes (polygalacturonases or pectate lyases) or bacteria lead to a smoothening of the surface and exposure of Alizarin according to Environmental Scanning Electron Microscopy (ESEM) analysis. Enzyme triggered release of Alizarin was demonstrated for a commercial enzyme preparation from Aspergillus niger and with purified polygalacturonase and pectate lyase from S. rolfsii and B. pumilus, respectively. In contrast to the PSA-CaCl(2) beads, cross-linking (PSA-GMA beads) restricted the release of Alizarin in absence of enzymes. There was a linear relation between release of Alizarin (5-348 μM) and enzyme activity in a range of 0-300 U ml(-1) dosed. In addition to enzymes, both PSA-CaCl(2) and PSA-GMA beads were incubated with Bacillus subtilis and Yersinia entercolitica as model contaminating microorganism. After 72 h, a release between 10 μM and 57 μM Alizarin was detected. For protection of the hydrogels, an enzymatically modified PET membrane was covalently attached onto the surface. This lead to a slower release and improve long term storage stability based on less than 1% release of dye after 21 days. Additionally, this allowed simple detection by visual inspection of the device due to a colour change of the white membrane to orange upon enzyme triggered release of the dye.
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor