MilliporeSigma
  • Plasma disposition, concentration in the hair, and anthelmintic efficacy of eprinomectin after topical administration in donkeys.
Plasma disposition, concentration in the hair, and anthelmintic efficacy of eprinomectin after topical administration in donkeys.
American journal of veterinary research (2011-12-01)
Cengiz Gokbulut, Antonio Di Loria, Necati Gunay, Roberto Masucci, Vincenzo Veneziano
ABSTRACT
To investigate plasma disposition, concentration in the hair, and anthelmintic efficacy of eprinomectin after topical administration in donkeys. 12 donkeys naturally infected with strongyle nematodes. The pour-on formulation of eprinomectin approved for use in cattle was administered topically to donkeys at a dosage of 0.5 mg/kg. Heparinized blood samples and hair samples were collected at various times between 1 hour and 40 days after administration. Samples were analyzed via high-performance liquid chromatography with fluorescence detection. Fecal strongyle egg counts were performed by use of a modified McMaster technique before and at weekly intervals for 8 weeks after treatment. Plasma concentration and systemic availability of eprinomectin were relatively higher in donkeys, compared with values reported for other animal species. Concerning the anthelmintic efficacy against strongyle nematodes, eprinomectin was completely effective (100%) on days 7 and 14 and highly effective (> 99%) until the end of the study at 56 days after treatment. No abnormal clinical signs or adverse reactions were observed for any donkeys after treatment. Eprinomectin had excellent safety. The relatively high plasma concentration after topical administration could result in use of eprinomectin for the control and treatment of parasitic diseases in donkeys.
Product Description
Supelco
Eprinomectin, PESTANAL®, analytical standard, mixture of isomers