MilliporeSigma
  • Gold-catalyzed formal [3 + 3] and [4 + 2] cycloaddition reactions of nitrosobenzenes with alkenylgold carbenoids.
Gold-catalyzed formal [3 + 3] and [4 + 2] cycloaddition reactions of nitrosobenzenes with alkenylgold carbenoids.
Journal of the American Chemical Society (2011-12-02)
Vinayak Vishnu Pagar, Appaso Mahadev Jadhav, Rai-Shung Liu
ABSTRACT
We report two new formal cycloaddition reactions between nitrosobenzenes and alkenylgold carbenoids. We obtained quinoline oxides 3 in satisfactory yields from the gold-catalyzed [3 + 3]-cycloadditions between nitrosobenzenes and alkenyldiazo esters 1. For propargyl esters 5, its resulting gold carbenes react with nitrosobenzene to give alkenylimine 8, followed by a [4 + 2]-cycloaddition with nitrosobenzene.
Sigma-Aldrich
Nitrosobenzene, ≥97%