MilliporeSigma
  • β-Cell-protective effect of 2-aminobicyclo-(2,2,1)-heptane-2-carboxylic acid as a glutamate dehydrogenase activator in db/db mice.
β-Cell-protective effect of 2-aminobicyclo-(2,2,1)-heptane-2-carboxylic acid as a glutamate dehydrogenase activator in db/db mice.
The Journal of endocrinology (2011-12-02)
Seung Jin Han, Sung-E Choi, Sang-A Yi, Soo-Jin Lee, Hae Jin Kim, Dae Jung Kim, Hyun Chul Lee, Kwan Woo Lee, Yup Kang
ABSTRACT
2-Aminobicyclo-(2,2,1)-heptane-2-carboxylic acid (BCH) is an activator of glutamate dehydrogenase (GDH), which is a mitochondrial enzyme with an important role in insulin secretion. We investigated the effect of BCH on the high-glucose (HG)-induced reduction in glucose-stimulated insulin secretion (GSIS), the HG/palmitate (PA)-induced reduction in insulin gene expression, and HG/PA-induced β-cell death. We also studied whether long-term treatment with BCH lowers blood glucose and improves β-cell integrity in db/db mice. We evaluated GSIS, insulin gene expression, and DNA fragmentation in INS-1 cells exposed to HG or HG/PA in the presence or absence of BCH. An in vivo study was performed in which 7-week-old diabetic db/db mice were treated with BCH (0.7  g/kg, n = 10) and placebo (n = 10) every other day for 6 weeks. After treatment, an intraperitoneal glucose tolerance test and immunohistological examinations were performed. Treatment with BCH blocked HG-induced GSIS inhibition and the HG/PA-induced reduction in insulin gene expression in INS-1 cells. In addition, BCH significantly reduced HG/PA-induced INS-1 cell death and phospho-JNK level. BCH treatment improved glucose tolerance and insulin secretion in db/db mice. BCH treatment also increased the ratio of insulin-positive β-cells to total islet area (P < 0.05) and reduced the percentage of β-cells expressing cleaved caspase 3 (P < 0.05). In conclusion, the GDH activator BCH improved glycemic control in db/db mice. This anti-diabetic effect may be associated with improved insulin secretion, preserved islet architecture, and reduced β-cell apoptosis.
Sigma-Aldrich
2-Amino-2-norbornanecarboxylic acid, amino acid transport inhibitor