MilliporeSigma
  • Unexpected anomeric selectivity of a 1-C-arylglycal donor in Kdo glycoside synthesis.
Unexpected anomeric selectivity of a 1-C-arylglycal donor in Kdo glycoside synthesis.
The Journal of organic chemistry (2011-12-07)
Yuxin Qian, Jianhao Feng, Masood Parvez, Chang-Chun Ling
ABSTRACT
A novel class of 1-C-arylglycals was developed and subjected to N-iodosuccinimide-mediated glycosylations with alcohols. Unexpectedly, all reactions provided 2-iodo-β-D-ketopyranosides in high yields and excellent stereoselectivity. After removal of the 2-iodide by radical conditions, the aryl group was smoothly oxidized to provide the corresponding β-Kdo glycosides. A mechanism for the stereoselective formation of β-D-ketopyranosides was proposed, which was supported by evidence from X-ray crystallography.
Sigma-Aldrich
N-Iodosuccinimide, 95%