MilliporeSigma
  • Eosinophilic esophagitis and pharyngitis presenting as mass lesion in a patient with inactive rheumatoid arthritis.
Eosinophilic esophagitis and pharyngitis presenting as mass lesion in a patient with inactive rheumatoid arthritis.
Journal of clinical rheumatology : practical reports on rheumatic & musculoskeletal diseases (2011-12-14)
Pulukool Sandhya, Debashish Danda, John Mathew, Susy Kurian, Balakrishna Siddhartha Ramakrishna
ABSTRACT
We describe here a case of longstanding rheumatoid arthritis (RA) presenting with recurrent episodes of epigastric pain, melena, nonprogressive dysphagia, and hoarseness associated with persistent peripheral blood eosinophilia. Her RA was clinically inactive, but she had significant lymphadenopathy and hepatosplenomegaly. Computed tomographic scan of the thorax revealed circumferential wall thickening extending from the oropharynx to the gastroesophageal junction with a large polypoidal mass projecting into the lumen of the stomach. Histology revealed infiltration of the esophageal mucosa by eosinophils with a density of 40 to 80 per high-power field. The stratified squamous epithelium of the pharyngeal mucosa was also infiltrated by eosinophils with a density of more than 100 per high-power field. Eosinophilic esophagitis and pharyngitis were diagnosed, and the patient was administered corticosteroids and hydroxyurea, following which her symptoms resolved. On repeat imaging, there was significant reduction in esophageal wall thickening and luminal dilatation. There are few reports of tissue eosinophilia in association with RA, but the pathogenesis and any definite association with RA are not clear.
Sigma-Aldrich
Deflazacort, ≥98% (HPLC)