MilliporeSigma
  • Synthesis and oligomerization of Fmoc/Boc-protected PNA monomers of 2,6-diaminopurine, 2-aminopurine and thymine.
Synthesis and oligomerization of Fmoc/Boc-protected PNA monomers of 2,6-diaminopurine, 2-aminopurine and thymine.
Organic & biomolecular chemistry (2011-12-14)
André H St Amant, Robert H E Hudson
ABSTRACT
A Boc-protecting group strategy for Fmoc-based PNA (peptide nucleic acid) oligomerization has been developed for thymine, 2,6-diaminopurine (DAP) and 2-aminopurine (2AP). The monomers may be used interchangeably with standard Fmoc PNA monomers. The DAP monomer was incorporated into a PNA and was found to selectively bind to T (ΔT(m)≥ +6 °C) in a complementary DNA strand. The 2AP monomer showed excellent discrimination of T (ΔT(m)≥ +12 °C) over the other nucleobases. 2AP also acted as a fluorescent probe of the PNA:DNA duplexes and displayed fluorescence quenching dependent on the opposite base.
Sigma-Aldrich
2,6-Diaminopurine, 98%