MilliporeSigma
  • Examining the feasibility of bulk commodity production in Escherichia coli.
Examining the feasibility of bulk commodity production in Escherichia coli.
Biotechnology letters (2011-12-14)
Claudia E Vickers, Daniel Klein-Marcuschamer, Jens O Krömer
ABSTRACT
Escherichia coli is currently used by many research institutions and companies around the world as a platform organism for the development of bio-based production processes for bulk biochemicals. A given bulk biochemical bioprocess must be economically competitive with current production routes. Ideally the viability of each bioprocess should be evaluated prior to commencing research, both by metabolic network analysis (to determine the maximum theoretical yield of a given biocatalyst) and by techno-economic analysis (TEA; to determine the conditions required to make the bioprocess cost-competitive). However, these steps are rarely performed. Here we examine theoretical yields and review available TEA for bulk biochemical production in E. coli. In addition, we examine fermentation feedstocks and review recent strain engineering approaches to achieve industrially-relevant production, using examples for which TEA has been performed: ethanol, poly-3-hydroxybutyrate, and 1,3-propanediol.
Sigma-Aldrich
1,3-Propanediol, 98%