MilliporeSigma
  • Effect of pullulan/poly(vinyl alcohol) blend system on the montmorillonite structure with property characterization of electrospun pullulan/poly(vinyl alcohol)/montmorillonite nanofibers.
Effect of pullulan/poly(vinyl alcohol) blend system on the montmorillonite structure with property characterization of electrospun pullulan/poly(vinyl alcohol)/montmorillonite nanofibers.
Journal of colloid and interface science (2011-12-17)
Md Shahidul Islam, Jeong Hyun Yeum, Ajoy Kumar Das
ABSTRACT
Nanofibers of the composite of pullulan (PULL), poly(vinyl alcohol) (PVA), and montmorillonite clay (MMT) were prepared using electrospinning method in aqueous solutions. Pullulan is an interesting natural polymer for many of its merits and good properties. Because of biocompatibility and non-toxicity of PVA, it could be used in numerous fields. Scanning electron microscopy (SEM), transmission electron microscopy (TEM), Fourier transform infrared (FTIR), X-ray diffraction (XRD), and thermal gravimetric analysis (TGA) were done to characterize the PULL/PVA/MMT nanofibers morphology and properties. XRD patterns and FTIR data demonstrated that there were good interactions between PULL and PVA caused by possibly hydrogen bonds. Moreover, XRD data and TEM images indicated that intercalated and exfoliated MMT nanoplatelets can be obtained within the PULL/PVA/MMT nanofibers depending on the PULL/PVA blend ratios. Furthermore, the thermal stability and mechanical property (tensile strength) of PULL/PVA/MMT nanofibers could be enhanced more by exfoliated MMT nanoplatelets than intercalated structures of that nanoplatelets.
Product Description
Supelco
Pullulan Standard Set Mp ~350 - 700′000, set of analytical standards, suitable for gel permeation chromatography (GPC)
Sigma-Aldrich
Pullulan, suitable for substrate for pullulanase