MilliporeSigma
  • Effect of feeding treatment during the backgrounding phase of beef production from pasture on: II. Longissimus muscle proximate composition, cholesterol and fatty acids.
Effect of feeding treatment during the backgrounding phase of beef production from pasture on: II. Longissimus muscle proximate composition, cholesterol and fatty acids.
Meat science (2011-12-24)
A J Pordomingo, T P García, G Volpi Lagreca
ABSTRACT
This study evaluated effects of feedlot backgrounding strategies (40, 70 or 100% alfalfa hay diets) or pasture grazing on moisture, protein, total lipids, ash, cholesterol concentration, and lipid profiles of the longissimus dorsi muscle (LM) of pasture finished Angus heifers. Ninety six calves were allocated to the strategies over a 114-day period, followed by pasture grazing over 132 days. At the end of the backgrounding stage, the concentration of omega-3 (n-3) fatty acids was highest in the Pasture group and this difference persisted (P<0.032) until the end of the 132 day pasture finishing phase. Similarly, the n-6/n-3 ratio was lowest in the Pasture group at the end of backgrounding and after pasture finishing. Backgrounding diets based on 70 and 100% hay or pasture grazing showed greater (P<0.041) conjugated linoleic acid (CLA) concentration in the lipid fraction than 40% hay. Results suggested that residual effects of backgrounding strategies could be detected in intramuscular fat of pasture finished heifers.
Product Description
Supelco
Conjugated (9E,11E)-Linoleic acid, analytical standard
Supelco
Conjugated (9Z,11E)-Linoleic acid, analytical standard